Assay Details
Target Gene Details
Entrez Gene ID: | 18534 |
Gene Name: | phosphoenolpyruvate carboxykinase 1, cytosolic |
Gene Aliases: |
AI265463, PEPCK, Pck-1 |
Location: |
Chr.2:173153073-173159254 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pck1 | NM_011044.2 | NP_035174.1 | ||
| AK028046.1 | BAC25718.1 | |||
| AK030327.1 | BAC26903.1 | |||
| AK133496.1 | BAE21687.1 | |||
| AK149525.1 | BAE28938.1 | |||
| BC037629.1 | AAH37629.1 |
More Information
Set Membership: |
|

Genomic Map