Assay Details
Target Gene Details
Entrez Gene ID: | 228983 |
Gene Name: | oxysterol binding protein-like 2 |
Gene Aliases: |
C130070J12Rik, ORP-2, Orp2, mKIAA0772 |
Location: |
Chr.2:180119366-180162680 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Osbpl2 | NM_144500.3 | NP_653083.2 | ||
| AK048537.1 | BAC33367.1 | |||
| AK145292.1 | BAE26349.1 | |||
| AK161695.1 | ||||
| AK166059.1 | ||||
| AK169504.1 | ||||
| AK170306.1 | BAE41703.1 | |||
| AK170669.1 | BAE41948.1 | |||
| AK178638.1 | ||||
| AK178810.1 | ||||
| AK182769.1 | ||||
| AK220257.1 | BAD90182.1 | |||
| BC026804.1 | AAH26804.1 |

Genomic Map