Assay Details
Target Gene Details
Entrez Gene ID: | 227720 |
Gene Name: | nucleoporin 214 |
Gene Aliases: |
BC039282, CAN, D2H9S46E |
Location: |
Chr.2:31974420-32053975 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup214 | NM_172268.2 | 3 | 434 | NP_758472.2 |
| AK122189.1 | 3 | 403 | BAC65471.1 | |
| AK150915.1 | 3 | 417 | BAE29953.1 | |
| AK166524.1 | 3 | 356 | BAE38828.1 | |
| BC039282.1 |
More Information
Set Membership: |
|

Genomic Map