Assay Details
Target Gene Details
Entrez Gene ID: | 18429 |
Gene Name: | oxytocin |
Gene Aliases: |
OT, Oxy |
Location: |
Chr.2:130574967-130577054 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxt | NM_011025.4 | NP_035155.1 | ||
| AK003042.1 | BAB22527.1 | |||
| BC117030.1 | ||||
| BC117032.1 |
More Information
Set Membership: |
|

Genomic Map