Assay Details
Target Gene Details
Entrez Gene ID: | 80708 |
Gene Name: | protein kinase C and casein kinase substrate in neurons 3 |
Gene Aliases: |
4921507A02Rik, 6330413E15Rik, AW413130 |
Location: |
Chr.2:91256098-91264680 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pacsin3 | NM_001289677.1 | 4 | 505 | NP_001276606.1 |
| NM_001289678.1 | 5 | 504 | NP_001276607.1 | |
| NM_028733.4 | 5 | 634 | NP_083009.1 | |
| NM_030880.3 | 5 | 500 | NP_112019.2 | |
| AF149824.1 | 5 | 418 | AAG31022.1 | |
| AF242531.1 | 4 | 403 | AAK29208.1 | |
| AK049166.1 | 5 | 635 | BAC33582.1 | |
| AK078060.1 | 4 | 513 | ||
| AK141063.1 | 5 | 477 | BAE24556.1 | |
| BC003884.1 | 5 | 441 | AAH03884.1 | |
| BY250979.1 |
More Information
Set Membership: |
|

Genomic Map