Assay Details
Target Gene Details
Entrez Gene ID: | 99035 |
Gene Name: | oleoyl-ACP hydrolase |
Gene Aliases: |
AI607300, E230009B14Rik, Thedc1 |
Location: |
Chr.2:3341982-3397238 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olah | NM_145921.1 | 2 | 267 | NP_666033.1 |
| AK087571.1 | 2 | 287 | BAC39932.1 | |
| AK136031.1 | 3 | 523 | BAE22784.1 | |
| BC025001.1 | 2 | 267 | AAH25001.1 |
More Information
Set Membership: |
|

Genomic Map