Assay Details
Target Gene Details
Entrez Gene ID: | 18610 |
Gene Name: | prodynorphin |
Gene Aliases: |
Dyn |
Location: |
Chr.2:129686549-129699938 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdyn | NM_001286502.1 | NP_001273431.1 | ||
| NM_018863.4 | NP_061351.2 | |||
| AF026537.1 | AAB82584.1 | |||
| AF483510.1 | AAL90784.1 | |||
| AF483511.1 | AAL90785.1 | |||
| AK133597.1 | BAE21743.1 | |||
| BC137571.1 | ||||
| BC137572.1 | ||||
| BY292284.1 |
More Information
Set Membership: |
|

Genomic Map