Assay Details
Target Gene Details
Entrez Gene ID: | 18549 |
Gene Name: | proprotein convertase subtilisin/kexin type 2 |
Gene Aliases: |
6330411F23Rik, AI839700, Nec-2, Nec2, PC2, Phpp-2, SPC2 |
Location: |
Chr.2:143546133-143816284 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcsk2 | NM_008792.4 | NP_032818.1 | ||
| AF483512.1 | AAL90786.1 | |||
| AF483513.1 | AAL90787.1 | |||
| AK133620.1 | BAE21752.1 | |||
| AK158493.1 | BAE34536.1 | |||
| AK158598.1 | BAE34574.1 | |||
| AK161585.1 | BAE36477.1 | |||
| BC052013.1 | AAH52013.2 | |||
| BC057348.1 | AAH57348.1 | |||
| M55669.1 | AAA39376.1 |
More Information
Set Membership: |
|

Genomic Map