Assay Details
Target Gene Details
Entrez Gene ID: | 18843 |
Gene Name: | BPI fold containing family A, member 1 |
Gene Aliases: |
LUNX, NASG, Plunc, SPLUNC1, SPURT |
Location: |
Chr.2:154142880-154149217 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifa1 | NM_011126.3 | NP_035256.2 | ||
| BC054375.1 | AAH54375.1 | |||
| U69172.1 | AAB63256.1 |

Genomic Map