Assay Details
Target Gene Details
Entrez Gene ID: | 66865 |
Gene Name: | peptidase (mitochondrial processing) alpha |
Gene Aliases: |
1200002L24Rik, 4933435E07Rik, Alpha-MPP, P-55 |
Location: |
Chr.2:26389348-26397121 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmpca | NM_173180.3 | NP_775272.1 | ||
| AK004549.1 | BAB23363.1 | |||
| AK032081.1 | ||||
| AK075965.1 | ||||
| AK077218.1 | BAC36690.1 | |||
| AK147110.1 | BAE27682.1 | |||
| AK150118.1 | BAE29321.1 | |||
| AK161291.1 | BAE36299.1 | |||
| AK167975.1 | ||||
| AK169325.1 | BAE41079.1 | |||
| AK169342.1 | BAE41094.1 |
Target Gene Details
Entrez Gene ID: | 68112 |
Gene Name: | serologically defined colon cancer antigen 3 |
Gene Aliases: |
C330016H24Rik, C630038K21Rik |
Location: |
Chr.2:26382798-26389316 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdccag3 | NM_001085407.1 | NP_001078876.1 | ||
| NM_001085408.1 | NP_001078877.1 | |||
| NM_026563.3 | NP_080839.2 | |||
| AK014885.1 | BAB29602.1 | |||
| AK021273.1 | BAB32357.1 | |||
| AK082787.1 | BAC38620.1 | |||
| AK163690.1 | BAE37459.1 | |||
| BC061037.1 | AAH61037.1 | |||
| BY104480.1 |
More Information
Set Membership: |
|

Genomic Map