Assay Details
Target Gene Details
Entrez Gene ID: | 66865 |
Gene Name: | peptidase (mitochondrial processing) alpha |
Gene Aliases: |
1200002L24Rik, 4933435E07Rik, Alpha-MPP, P-55 |
Location: |
Chr.2:26389348-26397121 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmpca | NM_173180.3 | NP_775272.1 | ||
| AK004549.1 | BAB23363.1 | |||
| AK032081.1 | ||||
| AK075965.1 | ||||
| AK077218.1 | BAC36690.1 | |||
| AK147110.1 | BAE27682.1 | |||
| AK150118.1 | BAE29321.1 | |||
| AK158990.1 | BAE34758.1 | |||
| AK161291.1 | BAE36299.1 | |||
| AK167975.1 | ||||
| AK169325.1 | BAE41079.1 | |||
| AK169342.1 | BAE41094.1 | |||
| BC010810.1 | AAH10810.1 |
More Information
Set Membership: |
|

Genomic Map