Assay Details
Target Gene Details
Entrez Gene ID: | 67996 |
Gene Name: | serine/arginine-rich splicing factor 6 |
Gene Aliases: |
1210001E11Rik, AI314910, AW146126, Sfrs6 |
Location: |
Chr.2:162931508-162937121 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf6 | NM_026499.4 | NP_080775.3 | ||
| AK004831.1 | BAB23599.1 | |||
| AK012605.1 | ||||
| AK133872.1 | BAE21902.1 | |||
| AK146293.1 | BAE27050.1 | |||
| AK159519.1 | BAE35148.1 | |||
| AK171709.1 | 2 | 402 | ||
| AK179065.1 | ||||
| AK188277.1 | ||||
| BC012039.1 | AAH12039.1 |
More Information
Set Membership: |
|

Genomic Map