Assay Details
Target Gene Details
Entrez Gene ID: | 228993 |
Gene Name: | solute carrier family 17, member 9 |
Gene Aliases: |
1700019H03Rik, BC019537, Vnut |
Location: |
Chr.2:180725213-180742280 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a9 | NM_183161.3 | NP_898984.3 | ||
| AK006122.1 | BAB24418.1 | |||
| BE850231.1 |
More Information
Set Membership: |
|

Genomic Map