Assay Details
Target Gene Details
Entrez Gene ID: | 228980 |
Gene Name: | TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
135kDa, AI450312, TAFII130, TAFII135, Taf2c1 |
Location: |
Chr.2:179912146-179976646 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf4a | NM_001081092.1 | NP_001074561.1 | ||
| AY038601.1 | AAK94779.1 |
More Information
Set Membership: |
|

Genomic Map