Assay Details
Target Gene Details
Entrez Gene ID: | 228684 |
Gene Name: | sel-1 suppressor of lin-12-like 2 (C. elegans) |
Gene Aliases: |
Gm118 |
Location: |
Chr.2:140229599-140389716 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sel1l2 | NM_001033296.2 | 15 | 1411 | NP_001028468.1 |
| AK016809.1 | 15 | 1357 | ||
| AK132650.1 | 15 | 1412 | BAE21280.1 | |
| BC150948.1 | 15 | 1383 |

Genomic Map