Assay Details
Target Gene Details
Entrez Gene ID: | 27219 |
Gene Name: | serum/glucocorticoid regulated kinase 2 |
Gene Aliases: |
AI098171, AW146006, Sgkl |
Location: |
Chr.2:162987302-163014139 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgk2 | NM_001291152.1 | 7 | 682 | NP_001278081.1 |
| NM_001291154.1 | 8 | 1401 | NP_001278083.1 | |
| NM_013731.3 | 8 | 800 | NP_038759.1 | |
| AF169033.1 | 8 | 735 | AAF12756.1 | |
| AK050009.1 | 8 | 1382 | BAC34031.1 | |
| AK136559.1 | 8 | 752 | BAE23044.1 | |
| BC026549.1 | 6 | 427 | AAH26549.1 |
More Information
Set Membership: |
|

Genomic Map