Assay Details
Target Gene Details
Entrez Gene ID: | 140476 |
Gene Name: | stereocilin |
Gene Aliases: |
DFNB16 |
Location: |
Chr.2:121363248-121381370 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strc | NM_080459.2 | NP_536707.2 | ||
| AF375593.1 | AAL35321.1 |
More Information
Set Membership: |
|

Genomic Map