Assay Details
Target Gene Details
Entrez Gene ID: | 19672 |
Gene Name: | reticulocalbin 1 |
Gene Aliases: |
Rcn |
Location: |
Chr.2:105385948-105399319 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcn1 | NM_009037.2 | 2 | 404 | NP_033063.1 |
| AK017494.1 | 2 | 311 | BAB30773.1 | |
| AK133971.1 | 2 | 354 | BAE21962.1 | |
| AK159973.1 | 2 | 307 | BAE35525.1 | |
| AK214949.1 | 1 | 36 | ||
| BC049108.1 | 2 | 297 | AAH49108.1 | |
| D13003.1 | 2 | 324 | BAA02366.1 |
More Information
Set Membership: |
|

Genomic Map