Assay Details
Target Gene Details
Entrez Gene ID: | 227656 |
Gene Name: | REX4, RNA exonuclease 4 homolog (S. cerevisiae) |
Gene Aliases: |
Gm111, Xpmc2h |
Location: |
Chr.2:26953563-26964539 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rexo4 | NM_207234.2 | NP_997117.2 | ||
| AB221675.1 | ||||
| AK045402.1 | 1 | 1066 | ||
| AK133982.1 | ||||
| AK138894.1 | BAE23812.1 | |||
| BC060147.1 | AAH60147.1 |
More Information
Set Membership: |
|

Genomic Map