Assay Details
Target Gene Details
Entrez Gene ID: | 56194 |
Gene Name: | PRP40 pre-mRNA processing factor 40 homolog A (yeast) |
Gene Aliases: |
2810012K09Rik, FBP11, Fnbp11, Fnbp3 |
Location: |
Chr.2:53134706-53191279 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf40a | NM_018785.2 | NP_061255.1 | ||
| AF135439.1 | AAD39463.1 |

Genomic Map