Assay Details
Target Gene Details
Entrez Gene ID: | 56194 |
Gene Name: | PRP40 pre-mRNA processing factor 40 homolog A (yeast) |
Gene Aliases: |
2810012K09Rik, FBP11, Fnbp11, Fnbp3 |
Location: |
Chr.2:53134706-53191279 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf40a | NM_018785.2 | 2 | 232 | NP_061255.1 |
| AF135439.1 | 2 | 232 | AAD39463.1 | |
| AK045207.1 | 2 | 250 | ||
| AK087963.1 | 2 | 149 | BAC40061.2 | |
| AK187647.1 | 2 | 99 | ||
| BC003318.1 | 2 | 259 |

Genomic Map