Assay Details
Target Gene Details
Entrez Gene ID: | 215653 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 2 |
Gene Aliases: |
3830431H01Rik, 9030412M04Rik, AI852669, AW495050 |
Location: |
Chr.2:131992850-132030287 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf2 | NM_175445.4 | 1 | 199 | NP_780654.1 |
| AK028384.1 | 1 | 103 | BAC25921.1 | |
| AK172906.1 | BAD32184.1 | |||
| BC057402.1 | AAH57402.1 | |||
| BQ770520.1 | 1 | 199 |
More Information
Set Membership: |
|

Genomic Map