Assay Details
Target Gene Details
Entrez Gene ID: | 110876 |
Gene Name: | sodium channel, voltage-gated, type II, alpha 1 |
Gene Aliases: |
6430408L10, A230052E19Rik, Nav1.2, Scn2a |
Location: |
Chr.2:65620764-65767449 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn2a1 | NM_001099298.2 | 18 | 3612 | NP_001092768.1 |
| AM905319.1 |
More Information
Set Membership: |
|

Genomic Map