Assay Details
Target Gene Details
Entrez Gene ID: | 68879 |
Gene Name: | PRP6 pre-mRNA splicing factor 6 homolog (yeast) |
Gene Aliases: |
1190003A07Rik, 2610031L17Rik, ANT-1, U5-102K |
Location: |
Chr.2:181601319-181655661 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf6 | NM_133701.2 | 1 | 59 | NP_598462.1 |
| AK011639.1 | BAB27751.1 | |||
| AK081998.1 | BAC38390.1 | |||
| AK145461.1 | 1 | 62 | BAE26451.1 | |
| AK166890.1 | ||||
| BC014869.1 | AAH14869.1 | |||
| BC023691.1 | AAH23691.2 | |||
| BC025030.1 | AAH25030.1 | |||
| BY735790.1 | 1 | 59 |
More Information
Set Membership: |
|

Genomic Map