Assay Details
Target Gene Details
Entrez Gene ID: | 51869 |
Gene Name: | Rap1 interacting factor 1 homolog (yeast) |
Gene Aliases: |
5730435J01Rik, 6530403D07Rik, AU016181, AW549474, D2Ertd145e |
Location: |
Chr.2:52072811-52126769 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rif1 | NM_175238.5 | NP_780447.4 | ||
| AK088688.1 | BAC40506.1 | |||
| AY428571.2 | AAR87833.2 | |||
| AY585206.1 | AAS94124.1 | |||
| JX255737.1 |
More Information
Set Membership: |
|

Genomic Map