Assay Details
Target Gene Details
Entrez Gene ID: | 20639 |
Gene Name: | U2 small nuclear ribonucleoprotein B |
Gene Aliases: |
2810052G09Rik |
Location: |
Chr.2:143063069-143072052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snrpb2 | NM_021335.3 | NP_067310.1 | ||
| AK005107.1 | BAB23823.1 | |||
| AK011274.1 | BAB27510.1 | |||
| AK013871.1 | BAB29026.1 | |||
| BC026794.1 | AAH26794.1 |
More Information
Set Membership: |
|

Genomic Map