Assay Details
Target Gene Details
Entrez Gene ID: | 20448 |
Gene Name: | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 |
Gene Aliases: |
SIAT7-D, ST6GalNAcIV, Siat7d |
Location: |
Chr.2:32587078-32599696 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6galnac4 | NM_001276425.1 | NP_001263354.1 | ||
| NM_011373.3 | NP_035503.1 | |||
| AJ007310.1 | CAA07446.1 | |||
| AK007601.1 | ||||
| AK085730.1 | BAC39523.1 | |||
| AK144884.1 | BAE26116.1 | |||
| AK153036.1 | BAE31668.1 | |||
| AK155956.1 | ||||
| AK159712.1 | BAE35308.1 | |||
| AK161559.1 | BAE36463.1 | |||
| AK179544.1 | ||||
| BC056451.1 | ||||
| Y15779.1 | CAB43507.1 | |||
| Y15780.1 | CAB43514.1 |

Genomic Map