Assay Details
Target Gene Details
Entrez Gene ID: | 50935 |
Gene Name: | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 |
Gene Aliases: |
ST6GalNAcVI, Siat7f |
Location: |
Chr.2:32599709-32620809 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6galnac6 | NM_001025311.2 | NP_001020482.1 | ||
| BY720739.1 |

Genomic Map