Assay Details
Target Gene Details
Entrez Gene ID: | 19730 |
Gene Name: | ral guanine nucleotide dissociation stimulator |
Gene Aliases: |
Gnds, Rgds, mKIAA1308 |
Location: |
Chr.2:28513089-28553082 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgds | NM_001145834.1 | NP_001139306.1 | ||
| NM_001145835.1 | NP_001139307.1 | |||
| NM_001145836.1 | NP_001139308.1 | |||
| NM_009058.2 | NP_033084.2 | |||
| AK129328.1 | BAC98138.1 | |||
| AK141959.1 | BAE24899.1 | |||
| AK149644.1 | BAE29002.1 | |||
| AK159020.1 | BAE34771.1 | |||
| AK159754.1 | BAE35345.1 | |||
| BC027225.1 | AAH27225.1 | |||
| L07924.1 | AAA37714.1 |
More Information
Set Membership: |
|

Genomic Map