Assay Details
Target Gene Details
Entrez Gene ID: | 20181 |
Gene Name: | retinoid X receptor alpha |
Gene Aliases: |
9530071D11Rik, Nr2b1, RXRalpha1 |
Location: |
Chr.2:27676324-27763319 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rxra | NM_001290481.1 | NP_001277410.1 | ||
| NM_001290482.1 | NP_001277411.1 | |||
| NM_011305.3 | NP_035435.1 | |||
| AK031034.1 | ||||
| AK144675.1 | BAE26004.1 | |||
| BC138800.1 | ||||
| BC138802.1 | ||||
| M84817.1 | AAA40080.1 | |||
| U77683.1 | AAB36777.1 | |||
| U77684.1 | AAB36778.1 | |||
| X66223.1 | CAA46962.1 |
More Information
Set Membership: |
|

Genomic Map