Assay Details
Target Gene Details
Entrez Gene ID: | 20648 |
Gene Name: | syntrophin, acidic 1 |
Gene Aliases: |
AW228934, Snt1 |
Location: |
Chr.2:154376314-154408084 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snta1 | NM_009228.2 | NP_033254.2 | ||
| AK137379.1 | BAE23334.1 | |||
| BC018546.1 | AAH18546.1 | |||
| U00677.1 | AAC52119.1 |

Genomic Map