Assay Details
Target Gene Details
Entrez Gene ID: | 50935 |
Gene Name: | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 |
Gene Aliases: |
ST6GalNAcVI, Siat7f |
Location: |
Chr.2:32599709-32620809 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6galnac6 | NM_001025310.2 | NP_001020481.1 | ||
| NM_001025311.2 | NP_001020482.1 | |||
| NM_001289547.1 | NP_001276476.1 | |||
| NM_001289548.1 | NP_001276477.1 | |||
| NM_001289549.1 | NP_001276478.1 | |||
| NM_016973.3 | NP_058669.1 | |||
| NR_110347.1 | ||||
| AB035123.1 | BAA95940.1 | |||
| AB035174.1 | BAA87036.1 | |||
| AK030648.1 | BAC27064.1 | |||
| AK157562.1 | ||||
| AK159158.1 | ||||
| AK207613.1 | ||||
| BC022924.1 |

Genomic Map