Assay Details
Target Gene Details
Entrez Gene ID: | 74030 |
Gene Name: | Ras and Rab interactor 2 |
Gene Aliases: |
2010003K16Rik, 4632403N06Rik, AW821980, RASSF4 |
Location: |
Chr.2:145771727-145887616 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rin2 | NM_028724.4 | NP_083000.5 | ||
| AK014548.1 | BAB29425.1 | |||
| AK040763.1 | BAC30697.1 | |||
| AK150680.1 | BAE29760.1 | |||
| AK151332.1 | BAE30311.1 | |||
| AK152025.1 | BAE30885.1 | |||
| AK152893.1 | BAE31576.1 | |||
| AK154421.1 | BAE32573.1 | |||
| AK163047.1 | BAE37170.1 | |||
| AK171064.1 | BAE42223.1 | |||
| AK198881.1 | ||||
| BC005529.1 | AAH05529.1 | |||
| BC040390.1 |
More Information
Set Membership: |
|

Genomic Map