Assay Details
Target Gene Details
Entrez Gene ID: | 74030 |
Gene Name: | Ras and Rab interactor 2 |
Gene Aliases: |
2010003K16Rik, 4632403N06Rik, AW821980, RASSF4 |
Location: |
Chr.2:145771727-145887616 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rin2 | NM_028724.4 | 6 | 651 | NP_083000.5 |
| AK150680.1 | 4 | 459 | BAE29760.1 | |
| AK151332.1 | 3 | 415 | BAE30311.1 | |
| AK152893.1 | 4 | 459 | BAE31576.1 | |
| BC040390.1 | 6 | 645 | ||
| CN461821.1 | 5 | 635 |
More Information
Set Membership: |
|

Genomic Map