Assay Details
Target Gene Details
Entrez Gene ID: | 215632 |
Gene Name: | pleckstrin and Sec7 domain containing 4 |
Gene Aliases: |
BC046518, EFA6B |
Location: |
Chr.2:24367561-24411223 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psd4 | NM_177611.3 | NP_808279.1 | ||
| AK043669.1 | BAC31612.1 | |||
| AK148489.1 | BAE28582.1 | |||
| AK150238.1 | BAE29402.1 | |||
| AK155249.1 | ||||
| AK163628.1 | BAE37428.1 | |||
| AK169859.1 | BAE41415.1 | |||
| BC046518.1 | AAH46518.1 | |||
| BC068231.1 | AAH68231.1 |
More Information
Set Membership: |
|

Genomic Map