Assay Details
Target Gene Details
Entrez Gene ID: | 378700 |
Gene Name: | BPI fold containing family B, member 3 |
Gene Aliases: |
Lplunc3, Rya3 |
Location: |
Chr.2:153918230-153932996 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifb3 | NM_194357.1 | NP_919338.2 | ||
| AK132627.1 | BAE21270.1 | |||
| BC048084.1 | AAH48084.2 |
More Information
Set Membership: |
|

Genomic Map