Assay Details
Target Gene Details
Entrez Gene ID: | 19383 |
Gene Name: | hnRNP-associated with lethal yellow |
Gene Aliases: |
AI663842, Merc |
Location: |
Chr.2:154791097-154871010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Raly | NM_001139511.1 | NP_001132983.1 | ||
| NM_001139512.1 | NP_001132984.1 | |||
| NM_001139513.1 | NP_001132985.1 | |||
| NM_023130.3 | NP_075619.2 | |||
| AK014356.1 | BAB29294.1 | |||
| AK135386.1 | BAE22515.1 | |||
| AK152290.1 | ||||
| AK154787.1 | BAE32830.1 | |||
| AK168781.1 | BAE40615.1 | |||
| BC004851.1 | AAH04851.1 | |||
| BC016587.1 | AAH16587.1 | |||
| L17076.1 | ||||
| S72641.1 | AAC60688.1 |
More Information
Set Membership: |
|

Genomic Map