Assay Details
Target Gene Details
Entrez Gene ID: | 378702 |
Gene Name: | small EDRK-rich factor 2 |
Gene Aliases: |
C80083, Msmac1l, m4F5rel |
Location: |
Chr.2:121449198-121453420 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serf2 | NM_001290837.1 | 1 | 98 | NP_001277766.1 |
| NM_011354.3 | 1 | 98 | NP_035484.1 | |
| NR_110986.1 | 1 | 98 | ||
| CJ073462.1 | 1 | 104 |

Genomic Map