Assay Details
Target Gene Details
Entrez Gene ID: | 26569 |
Gene Name: | solute carrier family 27 (fatty acid transporter), member 4 |
Gene Aliases: |
BB144259, FATP4 |
Location: |
Chr.2:29802680-29817522 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc27a4 | NM_011989.4 | 2 | 403 | NP_036119.1 |
| AJ251113.1 | 1 | 116 | CAC42082.1 | |
| AK036919.1 | 2 | 405 | BAC29639.1 | |
| AK155313.1 | 2 | 373 | ||
| AK155388.1 | 2 | 373 | BAE33236.1 | |
| AK170377.1 | 2 | 407 | BAE41756.1 | |
| AK194978.1 | ||||
| BC023114.1 | 2 | 254 | AAH23114.1 | |
| CT010312.1 | 1 | 116 | CAJ18520.1 |
More Information
Set Membership: |
|

Genomic Map