Assay Details
Target Gene Details
Entrez Gene ID: | 433470 |
Gene Name: | expressed sequence AA467197 |
Gene Aliases: |
Nmes1, mir-147 |
Location: |
Chr.2:122636971-122641087 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AA467197 |

Genomic Map