Assay Details
Target Gene Details
Entrez Gene ID: | 59029 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 14 |
Gene Aliases: |
2610312C03Rik, 3200001M20Rik, AA986732, Pad1, Poh1, rpm11 |
Location: |
Chr.2:61711694-61800376 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd14 | NM_021526.2 | NP_067501.2 | ||
| AK011974.1 | BAB27949.1 | |||
| AK012013.1 | BAB27974.1 | |||
| AK031863.1 | ||||
| AK078134.1 | ||||
| AK150280.1 | BAE29436.1 | |||
| AK151104.1 | BAE30114.1 | |||
| AK151733.1 | BAE30648.1 | |||
| BC003742.1 | AAH03742.1 | |||
| Y13071.1 | CAA73514.1 |
More Information
Set Membership: |
|

Genomic Map