Assay Details
Target Gene Details
Entrez Gene ID: | 68314 |
Gene Name: | hepatic nuclear factor 4 alpha, opposite strand |
Gene Aliases: |
0610008F07Rik |
Location: |
Chr.2:163493804-163541721 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnf4aos | NR_027970.1 | |||
| AK002328.1 | ||||
| BC025862.1 | AAH25862.1 |
Target Gene Details
Entrez Gene ID: | 100043899 |
Gene Name: | R3H domain containing-like |
Gene Aliases: |
OTTMUSG00000001070 |
Location: |
Chr.2:163491214-163504364 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| R3hdml | NM_001099331.2 | 4 | 742 | NP_001092801.1 |
More Information
Set Membership: |
|

Genomic Map