Assay Details
Target Gene Details
Entrez Gene ID: | 19334 |
Gene Name: | RAB22A, member RAS oncogene family |
Gene Aliases: |
3732413A17Rik, AI662177, AW319644, AW550514, E130120E14Rik, Rab22 |
Location: |
Chr.2:173659845-173702182 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab22a | NM_024436.3 | NP_077756.2 | ||
| AB232616.1 | BAF02878.1 | |||
| AJ311124.1 | CAC41378.1 | |||
| AK031667.1 | BAC27501.1 | |||
| AK146594.1 | BAE27289.1 | |||
| AK150617.1 | BAE29707.1 | |||
| AK219950.1 | ||||
| BC006596.1 | AAH06596.1 | |||
| M79304.1 | AAK14828.1 |

Genomic Map