Assay Details
Target Gene Details
Entrez Gene ID: | 574418 |
Gene Name: | serine incorporator 4 |
Gene Aliases: |
- |
Location: |
Chr.2:121451177-121456764 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serinc4 | NM_001025371.2 | NP_001020542.1 | ||
| AK044479.1 | BAC31945.1 | |||
| BC083132.1 | AAH83132.1 |

Genomic Map