Assay Details
Target Gene Details
Entrez Gene ID: | 378702 |
Gene Name: | small EDRK-rich factor 2 |
Gene Aliases: |
C80083, Msmac1l, m4F5rel |
Location: |
Chr.2:121449198-121453420 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serf2 | NM_001290837.1 | 3 | 2787 | NP_001277766.1 |
| NM_011354.3 | 3 | 2764 | NP_035484.1 | |
| NR_110986.1 | 3 | 2897 | ||
| AK028362.1 | 1 | 2273 | BAE20440.1 | |
| BP768998.1 | 1 | 338 |
Target Gene Details
Entrez Gene ID: | 574418 |
Gene Name: | serine incorporator 4 |
Gene Aliases: |
- |
Location: |
Chr.2:121451177-121456764 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serinc4 | NM_001025371.2 | NP_001020542.1 | ||
| AK044479.1 | BAC31945.1 | |||
| BC083132.1 | AAH83132.1 |
More Information
Set Membership: |
|

Genomic Map