Assay Details
Target Gene Details
Entrez Gene ID: | 68112 |
Gene Name: | serologically defined colon cancer antigen 3 |
Gene Aliases: |
C330016H24Rik, C630038K21Rik |
Location: |
Chr.2:26382798-26389316 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdccag3 | NM_001085407.1 | NP_001078876.1 | ||
| NM_001085408.1 | NP_001078877.1 | |||
| NM_026563.3 | NP_080839.2 | |||
| AK014885.1 | BAB29602.1 | |||
| AK021273.1 | BAB32357.1 | |||
| AK082787.1 | BAC38620.1 | |||
| AK163690.1 | BAE37459.1 | |||
| AW743523.1 | ||||
| BC031199.1 | AAH31199.1 | |||
| BC061037.1 | AAH61037.1 | |||
| BY104480.1 |

Genomic Map