Assay Details
Target Gene Details
Entrez Gene ID: | 70997 |
Gene Name: | sperm flagellar 1 |
Gene Aliases: |
4931426K16Rik, Clamp |
Location: |
Chr.2:131170261-131174810 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spef1 | NM_027641.2 | NP_081917.1 | ||
| AK016476.1 | ||||
| AK047675.1 | BAC33121.1 | |||
| AY860964.1 | AAX56299.1 | |||
| BC006046.1 | AAH06046.1 |

Genomic Map