Assay Details
Target Gene Details
Entrez Gene ID: | 108115 |
Gene Name: | solute carrier organic anion transporter family, member 4a1 |
Gene Aliases: |
OATP-E, Slc21a12 |
Location: |
Chr.2:180456250-180474853 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slco4a1 | NM_148933.1 | NP_683735.1 | ||
| AK033598.1 | BAC28379.1 | |||
| AK172688.1 | ||||
| BC030719.1 | AAH30719.1 | |||
| BC030720.1 | AAH30720.1 |
More Information
Set Membership: |
|

Genomic Map