Assay Details
Target Gene Details
Entrez Gene ID: | 20853 |
Gene Name: | staufen (RNA binding protein) homolog 1 (Drosophila) |
Gene Aliases: |
5830401L18Rik, AW549911, C85792, Stau |
Location: |
Chr.2:166947549-166996299 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stau1 | NM_001109905.2 | NP_001103375.1 | ||
| NM_001109906.2 | NP_001103376.1 | |||
| NM_001291160.1 | NP_001278089.1 | |||
| NM_011490.4 | NP_035620.1 | |||
| NR_111895.1 | ||||
| AF061942.1 | AAD17529.1 | |||
| AF395842.1 | AAK73752.1 | |||
| AK005002.2 | BAB23736.1 | |||
| AK139532.1 | BAE24052.1 | |||
| AK145826.1 | BAE26678.1 | |||
| AK151835.1 | BAE30728.1 | |||
| AK152269.1 | BAE31085.1 | |||
| AK152744.1 | BAE31464.1 | |||
| AK158952.1 | ||||
| AK159602.1 | BAE35221.1 | |||
| AK159781.1 | BAE35365.1 | |||
| AK160979.1 | BAE36128.1 | |||
| AK167637.1 | BAE39688.1 | |||
| AK194004.1 | ||||
| AK206904.1 | ||||
| AK210669.1 | ||||
| AY391773.1 | AAQ87664.1 | |||
| BC012959.1 | AAH12959.1 | |||
| BC082277.1 | AAH82277.1 |

Genomic Map