Assay Details
Target Gene Details
Entrez Gene ID: | 19194 |
Gene Name: | BPI fold containing family A, member 2 |
Gene Aliases: |
Bpifa2e, Psp |
Location: |
Chr.2:154008276-154016073 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifa2 | NM_008953.2 | NP_032979.1 | ||
| AK009032.1 | BAB26037.1 | |||
| AK009654.1 | BAB26418.1 | |||
| AK010181.1 | BAB26753.1 | |||
| BC010288.1 | AAH10288.1 | |||
| BC053336.1 | AAH53336.1 | |||
| X01697.1 | CAA25846.1 |

Genomic Map